56814 Fluka

Sodium phosphate dibasic

Ph Eur

DOWNLOAD MSDS (PDF)

Synonym: sec-Sodium phosphate, Dinatrii phosphas anhydricus, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

  • CAS Number 7558-79-4

  • Linear Formula Na2HPO4

  • Molecular Weight 141.96

  •  EC Number 231-448-7

  •  MDL number MFCD00003496

Properties

Related Categories Chemical Synthesis, Inorganic Salts, Sodium, Synthetic Reagents
grade   Ph Eur
pharmacopeia   testing & handling conforms to Pharmacopeia
pKa (25 °C)   (1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

Price and Availability

Customers Also Viewed

Potassium phosphate monobasic

Ph Eur, anhydrous

Sodium phosphate dibasic

puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)

Sodium phosphate dibasic

puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)

Sodium phosphate dibasic

purum p.a., anhydrous, ≥98.0% (T)

Sodium phosphate dibasic heptahydrate

meets USP testing specifications

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?