CAS Number 607-80-7
Empirical Formula (Hill Notation) C20H18O6
Molecular Weight 354.35
MDL number MFCD00216134
| Related Categories | Analytical Standards, Analytical/Chromatography, Artemisia, Chromatography, Life Sciences Standards, |
| grade | analytical standard |
| assay | ≥98.0% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
| storage temp. | 2-8°C |
Non-competitive Δ5-desaturase inhibitor.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?