CAS Number 3811-04-9
Linear Formula KClO3
Molecular Weight 122.55
EC Number 223-289-7
MDL number MFCD00011361
Popular Documents: FTIR (PDF)
| Related Categories | Chemical Synthesis, Inorganic Salts, Potassium, Synthetic Reagents |
| grade | purum p.a. |
| assay | ≥99.0% (T) |
| quality | crystallized |
| mp | 356 °C(lit.) |
| anion traces | chloride (Cl-): ≤50 mg/kg |
| sulfate (SO42-): ≤50 mg/kg | |
| cation traces | Ca: ≤50 mg/kg |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?