60415 Fluka

Potassium nitrate

purum p.a., ≥98.0% (T)

DOWNLOAD MSDS (PDF)
  • CAS Number 7757-79-1

  • Empirical Formula (Hill Notation) KNO3

  • Molecular Weight 101.10

  •  EC Number 231-818-8

  •  MDL number MFCD00011409

Properties

Related Categories Chemical Synthesis, Inorganic Salts, Potassium, Synthetic Reagents
grade   purum p.a.
assay   ≥98.0% (T)
impurities   ≤1.0% anticaking agent (Tixosil)
mp   334 °C(lit.)
anion traces   chloride (Cl-): ≤500 mg/kg
  sulfate (SO42-): ≤200 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  Na: ≤1000 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Description

Analysis Note

anticaking agent added

Price and Availability

Customers Also Viewed

Potassium nitrate

BioXtra, ≥99.0%

Potassium nitrate

TraceSELECT®, for trace analysis, ≥99.995% (metals basis)

Potassium nitrate

ACS reagent, ≥99.0%

Potassium nitrate

ReagentPlus®, ≥99.0%

Potassium nitrate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?