61520 Fluka

Lanthanum(III) nitrate hexahydrate

puriss. p.a., ≥99.0% (titration)

DOWNLOAD MSDS (PDF)

Synonym: Nitric acid, lanthanum (III) salt, hexahydrate

  • CAS Number 10277-43-7

  • Linear Formula La(NO3)3 · 6H2O

  • Molecular Weight 433.01

  •  EC Number 233-238-0

  •  MDL number MFCD00149751

  •  PubChem Substance ID 24882149

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Catalysis and Inorganic Chemistry, Chemical Synthesis,
grade   puriss. p.a.
assay   ≥99.0% (titration)
form   crystalline
mp   65-68 °C
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤10 mg/kg
  K: ≤10 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤10 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤10 mg/kg
  Zn: ≤5 mg/kg

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Cerium(III) nitrate hexahydrate

99.999% trace metals basis

Cerium(III) nitrate hexahydrate

puriss. p.a., ≥99.0%

Lanthanum nitrate solution

volumetric, 0.1 M La(NO3)3, for the determination of fluoride

Lanthanum(III) nitrate hexahydrate

99.999% trace metals basis

Lanthanum(III) nitrate hexahydrate

99.99% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?