Synonym: Lauric acid methyl ester, Methyl laurate
CAS Number 111-82-0
Linear Formula CH3(CH2)10COOCH3
Molecular Weight 214.34
Beilstein Registry Number 1767780
EC Number 203-911-3
MDL number MFCD00008966
PubChem Substance ID 24874824
Popular Documents: FTNMR (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Biochemicals and Reagents, Chromatography, |
| grade | analytical standard |
| assay | ≥99.5% (GC) |
| shelf life | (limited shelf life, expiry date on the label) |
| refractive index |
n |
| bp | 261-262 °C(lit.) |
| mp | 4.5-5 °C |
| density | 0.869 g/mL at 20 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?