Synonym:
α-Lnn,
cis,cis,cis-
CAS Number 463-40-1
Linear Formula CH3(CH2CH=CH)3(CH2)7CO2H
Molecular Weight 278.43
Beilstein Registry Number 1727693
EC Number 207-334-8
MDL number MFCD00065720
PubChem Substance ID 24882198
Popular Documents: FTNMR (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Applications, Biochemicals and Reagents, |
| grade | analytical standard |
| assay | ≥98.5% (GC) |
| shelf life | (limited shelf life, expiry date on the label) |
| refractive index |
n |
|
n | |
| bp | 230-232 °C/1 mmHg(lit.) |
| mp | -11 °C(lit.) |
An ω-3 fatty acid that serves as a precursor to eicosapentaenoic acid (EPA) but not docosahexaenoic acid. Conversion is greater in women than men, and conversely, β-oxidation metabolism is greater in men than women.1
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?