Synonym:
2-
CAS Number 132299-21-9
Empirical Formula (Hill Notation) C21H14K4N2O9
Molecular Weight 594.74
MDL number MFCD00133417
PubChem Substance ID 24882285
| grade | for fluorescence |
| storage temp. | 2-8°C |
Fluorescent Mg2+ indicator.1
Fluorescent Mg2+ indicator, useful for the study of intracellular Mg2+ concentrations1,2
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?