Synonym: Propanedioic acid
CAS Number 141-82-2
Linear Formula CH2(COOH)2
Molecular Weight 104.06
Beilstein Registry Number 1751370
MDL number MFCD00002707
| Related Categories | Acid, Acids and Esters, All Alphabetically Sorted, Analytical Standards, Analytical/Chromatography, |
| grade | certified reference material |
| TraceCERT® | |
| shelf life | (limited shelf life, expiry date on the label) |
| mp | 132-135 °C (dec.)(lit.) |
Certified content by quantitative NMR incl. uncertainty and expiry are given in the certificate.
Download your certificate at: http://www.sigma-aldrich.
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?