Synonym: NOGE 3-Ring (Novolac glycidyl ether) mixture of isomers, Novolac glycidyl ether 3-Ring
CAS Number 158163-01-0
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, Food & Beverage Standards, |
| grade | analytical standard |
| assay | ≥90.0% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
| storage temp. | 2-8°C |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?