70136 Fluka

Deoxyribonuclease Test Agar

for microbiology

DOWNLOAD MSDS (PDF)

Synonym: DNase Test Agar

Properties

Related Categories A - M, All Food Control Media (alphabetical), All Solid Media/Agars, Biochemical Identification Media by Test, Biochemical Identification Media, A to Z,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
composition   agar, 15 g/L
  casein enzymic hydrolysate, 15 g/L
  deoxyribonucleic acid, 2 g/L
  papaic digest of soybean meal, 5 g/L
  sodium chloride, 5 g/L
final pH   7.3±0.2 (25 °C)
suitability   nonselective and differential for Serratia
  nonselective and differential for Staphylococcus
  nonselective and differential for Streptococcus

Description

Application

A diagnostic medium for the detection of coagulase-positive DNase producing Staphylococci.

Other Notes

In an identification scheme for coagulase negative staphylococci from bovine mastitis1

Price and Availability

Customers Also Viewed

Deoxyribonuclease I bovine

recombinant, expressed in Pichia pastoris, buffered aqueous glycerol solution, ≥5,000 units/mg protein

for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?