70148 Fluka

Nutrient Agar

for microbiology

DOWNLOAD MSDS (PDF)

Properties

Related Categories All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Solid Media/Agars, Aspergillus, Bacillus,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
composition   agar, 15 g/L
  meat extract, 1 g/L
  peptone, 5 g/L
  sodium chloride, 5 g/L
  yeast extract, 2 g/L
final pH   7.1±0.2 (25 °C)
suitability   nonselective for Aspergillus
  nonselective for Bacillus
  nonselective for Bacteria (General Media)
  nonselective for Candida
  nonselective for Enterococcus
  nonselective for Escherichia coli and Coliforms
  nonselective for Listeria
  nonselective for Pseudomonas
  nonselective for Staphylococcus
  nonselective for Streptococcus

Description

Application

A general culture medium for less fastidious microorganisms as well as for permanent cultures. Add blood serum, or other biological fluids if required.

Other Notes

Studies on heterotrophic bacteria in some Argentine soils1

Price and Availability

Customers Also Viewed

for microbiology

Microbiologically tested

Microbiologically tested

for microbiology

No. 1, for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?