Synonym: Standard - Nutrient Broth
MDL number MFCD01867738
Popular Documents: Datasheet (PDF)
| Related Categories | All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Liquid Media/Broths, Bacteria (General Media), Clinical Media, |
| grade | for microbiology |
| shelf life | (limited shelf life, expiry date on the label) |
| composition | meat extract, 1 g/L |
| peptone, 5 g/L | |
| sodium chloride, 5 g/L | |
| yeast extract, 2 g/L | |
| final pH | 7.4±0.2 (25 °C) |
An inexpensive liquid medium which serves as base for the culture and accumulation of microorganisms.
Customers Also Viewed
for microbiology
Microbiologically tested
No. 1, for microbiology
No. 2, for microbiology
No. 4, for microbiology
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?