70166 Fluka

Skim Milk Powder

for microbiology

DOWNLOAD MSDS (PDF)

Synonym: Milk powder fat-free

Properties

Related Categories Base Ingredients for Media, Microbiology, Other Additives, Protein Sources (Peptones & Extracts), Protein Sources (milk derived),
grade   for microbiology
availability   not available in USA and Canada
impurities   ≤1.5% lipid
  ≤5.0% water
  ~5.3% total nitrogen (N)
  ~55% reducing sugars (as lactose monohydrate)
ign. residue   ≤10%

Description

Other Notes

Sales restrictions may apply

Reconstitution

Mix the powder to a smooth paste with a small quantity of distilled water, then gradually add more distilled water until a 10% w/v mixture is obtained. This is equivalent to fresh milk, and may be sterilized by autoclaving for 5 minutes at 121 °C. Care should be taken not to overheat during sterilization, otherwise caramelization will occur.

Price and Availability

Customers Also Viewed

with 5% nonfat milk, powder (dry milled), pH 7.3

for microbiology

BCR® certified Reference Material

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?