70191 Fluka

Mueller Hinton Agar

for microbiology

DOWNLOAD MSDS (PDF)

Properties

Related Categories A - M, All Solid Media/Agars, Bacteria (General Media), Clinical Media, Media,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
composition   agar, 17.0 g/L
  beef infusion solids, 2.0 g/L
  casein hydrolysate, 17.5 g/L
  starch, 1.5 g/L
final pH   7.3±0.2 (25 °C)
suitability   nonselective for Bacteria (General Media)
  nonselective for Neisseria

Description

Application

A solid medium originally designed for the isolation of pathogenic Neisseria species, now widely used for antibiotic susceptibility testing (including sulfonamides).

Other Notes

Enhancement of cefotoxamine and other cephalosporins against Enterococcus faecalis by blood supplemented Mueller-Hinton agar1,2

Price and Availability

Customers Also Viewed

for microbiology

for microbiology

for microbiology

for microbiology, Cation-adjusted

for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?