74280 Fluka

1-Nonanol

purum, ≥98.0% (GC)

DOWNLOAD MSDS (PDF)

Synonym: Alcohol C9, Nonyl alcohol

  • CAS Number 143-08-8

  • Linear Formula CH3(CH2)8OH

  • Molecular Weight 144.25

  •  Beilstein Registry Number 969213

  •  EC Number 205-583-7

  •  MDL number MFCD00002990

  •  PubChem Substance ID 24886672

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alcohols, Biochemicals and Reagents, Building Blocks, C9 to C10, Chemical Synthesis,
vapor density   5 (vs air)
vapor pressure   13 mmHg ( 104 °C)
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.433(lit.)
  n20/D 1.434
bp   215 °C(lit.)
mp   −8-−6 °C(lit.)
density   0.827 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

1-Nonanol
1312101-Nonanol

98%

1-Undecanol

99%

Nonyl alcohol

≥98%, FCC

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?