74850 Fluka

1-Octanol

puriss., ≥99.5% (GC)

DOWNLOAD MSDS (PDF)

Synonym: Alcohol C8, Capryl alcohol, Octyl alcohol

  • CAS Number 111-87-5

  • Linear Formula CH3(CH2)7OH

  • Molecular Weight 130.23

  •  Beilstein Registry Number 1697461

  •  EC Number 203-917-6

  •  MDL number MFCD00002988

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alcohols, Aspalathus linearis (Rooibos tea), Biochemicals and Reagents, Boswellia carterii, Building Blocks,
vapor density   4.5 (vs air)
vapor pressure   0.14 mmHg ( 25 °C)
grade   puriss.
assay   ≥99.5% (GC)
autoignition temp.   523 °F
refractive index   n20/D 1.429(lit.)
  n20/D 1.429
bp   196 °C(lit.)
mp   −15 °C(lit.)
density   0.827 g/mL at 25 °C(lit.)

Description

Biochem/physiol Actions

Blocks T-type Ca2+ channels

Price and Availability

Customers Also Viewed

1-Octanol

analytical standard

1-Octanol

≥99.5% (GC)

1-Octanol
2978871-Octanol

anhydrous, ≥99%

1-Octanol
2932451-Octanol

CHROMASOLV®, for HPLC, ≥99%

1-Octanol
1126151-Octanol

ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?