Synonym: 1-Octanesulfonic acid sodium salt
CAS Number 5324-84-5
Linear Formula CH3(CH2)6CH2SO3Na
Molecular Weight 216.27
Beilstein Registry Number 3574241
EC Number 231-791-2
MDL number MFCD00007544
PubChem Substance ID 24886749
Popular Documents: FTIR (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, Basic Ion Pair Reagents (Cationic), Buffers & Reagents, Chromatography Reagents & Solvents, |
| grade | for ion pair chromatography |
| quality | ampule |
| λ | in concentrate |
| UV absorption | λ: 210 nm Amax: 0.08 |
| λ: 220 nm Amax: 0.05 | |
| λ: 230 nm Amax: 0.03 | |
| λ: 260 nm Amax: 0.02 |
concentrate (~0.33 M) in ampules for 6x1 l 0.005 M ready-to-use solution
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?