Empirical Formula (Hill Notation) C28H37F3O4
Molecular Weight 494.59
MDL number MFCD01863101
PubChem Substance ID 24886799
| Related Categories | Biochemicals and Reagents, Detection, Fluorescent Enzyme Substrates, Fluorescent Enzyme Substrates for Spectroscopy, Fluorescent Probes, Labels, Particles and Stains More... |
| grade | for fluorescence |
| solubility | DMSO: soluble |
| dioxane: soluble | |
| fluorescence | λex 385 nm; λem 502 nm in 0.1 M Tris pH 8.0 (lipase) |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?