Synonym:
1-
CAS Number 1231710-62-5
Empirical Formula (Hill Notation) C11D3H12NO2 · HCl
Molecular Weight 232.72
MDL number MFCD18910754
| Related Categories | Alkaloids, Amphetamine, Analytical Standards, Analytical/Chromatography, Chemical Structure, |
| grade | analytical standard, for drug analysis |
| assay | ≥98.5% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
| mass shift | M+3 |
| storage temp. | 2-8°C |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?