| Related Categories | Analytical/Chromatography, GC Stationary Phases, Gas Chromatography (GC), Packed GC Columns and Components |
| grade | for GC |
| packaging | bottle of 100 mL |
| refractive index |
n |
| matrix active group | DC-710 phase |
| viscosity | ~500 mPa.s, neat(25 °C) |
Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.
Customers Also Viewed
for biotechnological purposes
viscosity ~50 mPa.s, neat(25 °C)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?