86853 Fluka

Tetrabutylammonium bisulfate

for ion pair chromatography, ≥99.0%

DOWNLOAD MSDS (PDF)

Synonym: Tetrabutylammonium hydrogen sulfate

  • CAS Number 32503-27-8

  • Linear Formula (CH3CH2CH2CH2)4N(HSO4)

  • Molecular Weight 339.53

  •  Beilstein Registry Number 3599663

  •  EC Number 251-068-5

  •  MDL number MFCD00011637

  •  PubChem Substance ID 24888782

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Acidic Ion Pair Reagents (Anionic), Analytical Reagents, Analytical/Chromatography, Buffers & Reagents, Chromatography Reagents & Solvents,
grade   for ion pair chromatography
assay   ≥99.0% (T)
  ≥99.0%
mp   171-174 °C
anion traces   bromide (Br-): ≤50 mg/kg
λ   10 % in H2O
UV absorption   λ: 210 nm Amax: 0.06
  λ: 220 nm Amax: 0.05
  λ: 230 nm Amax: 0.03
  λ: 260 nm Amax: 0.02
  λ: 500 nm Amax: 0.02
suitability   corresponds to standard for RP gradient test
  corresponds to standard for filter test

Price and Availability

Customers Also Viewed

Tetrabutylammonium bisulfate

puriss., ≥99.0% (T)

Tetrabutylammonium bisulfate

purum, ≥97.0% (T)

Tetrabutylammonium bisulfate solution

for ion pair chromatography, concentrate, ampule

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?