Synonym: Tetrapropylammonium hydrogen sulfate
CAS Number 56211-70-2
Empirical Formula (Hill Notation) C12H29NO4S
Molecular Weight 283.43
Beilstein Registry Number 6833419
MDL number MFCD00038764
PubChem Substance ID 24889054
Popular Documents: RAMAN FTIR (PDF)
| Related Categories | Acidic Ion Pair Reagents (Anionic), Analytical Reagents, Analytical/Chromatography, Buffers & Reagents, Chromatography Reagents & Solvents, |
| grade | for ion pair chromatography |
| assay | ≥99.0% (T) |
| ≥99.0% | |
| mp | 158-160 °C |
| λ | 10 % in H2O |
| UV absorption | λ: 210 nm Amax: 0.05 |
| λ: 220 nm Amax: 0.04 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?