91366 Fluka

Universal Pre-Enrichment Broth

for microbiology

DOWNLOAD MSDS (PDF)

Properties

Related Categories All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Liquid Media/Broths, Listeria, Media,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
composition   casein enzymic hydrolysate, 5.0 g/L
  dextrose, 0.50 g/L
  disodium phosphate, 7.0 g/L
  ferric ammonium citrate, 0.10 g/L
  magnesium sulfate, 0.250 g/L
  monopotassium phosphate, 15.0 g/L
  proteose peptone, 5.0 g/L
  sodium chloride, 5.0 g/L
  sodium pyruvate, 0.20 g/L
final pH   6.3±0.2 (25 °C)
suitability   nonselective for Listeria
  nonselective for Salmonella

Description

Application

Universal Pre-enrichment Broth is used for recovering sublethally injured Salmonella and Listeria from food products.

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?