Synonym:
Eu(hfc)3,
Tris[3-
CAS Number 34788-82-4
Empirical Formula (Hill Notation) C42H42EuF21O6
Molecular Weight 1193.71
EC Number 252-214-0
MDL number MFCD00151059
PubChem Substance ID 24889844
Popular Documents: FTIR (PDF)
| Related Categories | Asymmetric Synthesis, Chemical Synthesis, Chiral Resolution Reagents, Chiral Shift Reagents |
| grade | for NMR spectroscopy |
| puriss. p.a. | |
| mp | ~160 °C(lit.) |
| storage temp. | 2-8°C |
Chiral shift reagent for the determination of enantiomer ratios by NMR 1,2; Measurements in polar solvents3; For use as a Lanthanide shift reagent4
1 g in glass bottle
250 mg in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?