93560 Fluka

Tropylium tetrafluoroborate

purum, ≥97.0% (T)

DOWNLOAD MSDS (PDF)

Synonym: Cycloheptatrienyl tetrafluoroborate, Tropylium fluoroborate

  • CAS Number 27081-10-3

  • Empirical Formula (Hill Notation) C7H7BF4

  • Molecular Weight 177.94

  •  Beilstein Registry Number 3574512

  •  EC Number 248-214-5

  •  MDL number MFCD00013402

Properties

Related Categories Building Blocks, Carbenium Salts, Chemical Synthesis, Organic Building Blocks
grade   purum
assay   ≥97.0% (T)
mp   ~240 °C (dec.)(lit.)

Description

Application

Reactant or reagent involved in:
• Retro-Buchner reactions for synthesis of arylcyclopropanes1
• Desymmetrization for access to homocalystegine analogs2 or aminocycloheptitols3
• α-Cyanation of imines4
• Stereoselective α-alkylation of aldehydes with stable carbocations5
• Oxidative C-H activation of aliphatic aldehydes6
• Electrochemical Redox transformations7

Price and Availability

Customers Also Viewed

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?