94425 Fluka

Propionic acid

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: Acid C3, Propanoic acid, Propanyl acid

  • CAS Number 79-09-4

  • Linear Formula CH3CH2COOH

  • Molecular Weight 74.08

  •  Beilstein Registry Number 506071

  •  EC Number 201-176-3

  •  MDL number MFCD00002756

  •  PubChem Substance ID 24889994

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Acid, All Alphabetically Sorted, Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards,
grade   analytical standard
vapor density   2.55 (vs air)
vapor pressure   2.4 mmHg ( 20 °C)
assay   ≥99.8% (GC)
autoignition temp.   955 °F
shelf life   (limited shelf life, expiry date on the label)
expl. lim.   12.1 %
refractive index   n20/D 1.386(lit.)
  n20/D 1.386
bp   141 °C(lit.)
mp   −24-−23 °C(lit.)
density   0.993 g/mL at 25 °C(lit.)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

Acetic acid

analytical standard

Butyric acid

analytical standard

Propionic acid

ACS reagent, ≥99.5%

Propionic acid

≥99.5%

Propionic acid

BioReagent, suitable for insect cell culture, ~99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?