95586 Fluka

XLD Agar

for microbiology

DOWNLOAD MSDS (PDF)

Synonym: Xylose Lysine Deoxycholate Agar

Properties

Related Categories All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Solid Media/Agars, Enterobacteriaceae, Klebsiella,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
composition   agar, 15 g/L
  ammonium ferric citrate, 0.8 g/L
  lactose, 7.5 g/L
  L-lysine hydrochloride, 5 g/L
  phenol red, 0.08 g/L
  sodium chloride, 5 g/L
  sodium deoxycholate, 2.5 g/L
  sodium thiosulfate, 6.8 g/L
  sucrose, 7.5 g/L
  xylose, 3.5 g/L
  yeast extract, 3 g/L
final pH   7.4±0.2 (25 °C)
suitability   selective and differential for Enterobacteriaceae
  selective and differential for Klebsiella
  selective and differential for Proteus
  selective and differential for Salmonella
  selective and differential for Shigella

Description

Application

For the isolation and differentiation of pathogenic enterobacteriaceae in particular the species Shigella and Salmonella acc. to Taylor (1965), Taylor and Harris (1965, 1967) and Taylor and Schelhart (1967).

Price and Availability

Customers Also Viewed

85640SS-Agar

for microbiology

for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?