CAS Number 51-63-8
Empirical Formula (Hill Notation) C9H13N · 0.5 H2SO4
Molecular Weight 184.25
MDL number MFCD00069209
PubChem Substance ID 24890756
Popular Documents: Datasheet (PDF) | Specification Sheet (PDF)
| Related Categories | β-Glucuronidase, β-Glucuronidase Drug Analysis Bundle, A, AM to AQ, Alphabetical Index of Analytical Standards, |
| grade | analytical standard, for drug analysis |
| drug control | Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschland |
| concentration | 1.0 mg/mL±5% in methanol |
| storage temp. | 2-8°C |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?