Synonym: MS-222, Tricaine
CAS Number 886-86-2
Empirical Formula (Hill Notation) C9H11NO2 · CH4SO3
Molecular Weight 261.29
Beilstein Registry Number 3775022
EC Number 212-956-8
MDL number MFCD00013176
PubChem Substance ID 24890935
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, E, EQ - EZ More... |
| mp | ~148 °C |
25, 100, 250 g in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?