Synonym: α-(1-[t-Butylamino]ethyl)-2,5-dimethoxybenzyl alcohol
CAS Number 5696-15-1
Empirical Formula (Hill Notation) C15H25NO3 · HCl
Molecular Weight 303.82
EC Number 227-169-5
MDL number MFCD00135781
PubChem Substance ID 24891587
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, B, BI - BZ, |
| grade | analytical standard, for drug analysis |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?