Synonym: Mineral oil
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Application CRMs, Certified Reference Materials (CRMs), |
| grade | BCR® certified Reference Material |
BCR is a registered trademark of European Commission
Customers Also Viewed
BioReagent, for molecular biology, light oil
PCR Reagent
meets USP testing specifications
BCR® certified Reference Material, low level
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?