Synonym:
5-
CAS Number 22494-42-4
Empirical Formula (Hill Notation) C13H8F2O3
Molecular Weight 250.20
EC Number 245-034-9
MDL number MFCD00057834
PubChem Substance ID 24893742
Popular Documents: Specification Sheet (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Bioactive Small Molecules, Cell Biology, |
| grade | analytical standard |
| Gene Information | human ... ALB(213) |
Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. D3281.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?