E5399 Fluka

Endo agar

for microbiology

DOWNLOAD MSDS (PDF)

Properties

Related Categories A - M, All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Solid Media/Agars, Enterobacter,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
pH   7.5±0.2 (25 °C)
suitability   selective and differential for Enterobacter
  selective and differential for Escherichia coli and Coliforms
  selective and differential for Klebsiella
  selective and differential for Proteus
  selective and differential for Salmonella
  selective and differential for Shigella

Description

Components

Components (g/L)
Peptic Digest of Animal Tissue 10.00
Lactose 10.00
Dipotassium Phosphate 3.50
Sodium Sulphite 2.50
Basic Fuchsin 0.50
Agar 15.00

Application

Recommended for the detection of coliform and other enteric organisms.

Price and Availability

Customers Also Viewed

A7002Agar

ash 2.0-4.5%

A1296Agar

microbiology tested, plant cell culture tested, cell culture tested, powder

for molecular biology

for microbiology

for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?