H5149 Fluka

Heneicosanoic acid

analytical standard

DOWNLOAD MSDS (PDF)
  • CAS Number 2363-71-5

  • Linear Formula CH3(CH2)19COOH

  • Molecular Weight 326.56

  •  Beilstein Registry Number 1711889

  •  EC Number 219-113-3

  •  MDL number MFCD00002805

  •  PubChem Substance ID 24895676

Properties

Related Categories Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, H, HA -HT More...
grade   analytical standard
assay   ≥99% (capillary GC)
mp   74-75 °C(lit.)
storage temp.   2-8°C

Description

Packaging

1 g in poly bottle

100 mg in poly bottle

Price and Availability

Customers Also Viewed

Heneicosanoic acid

purum, ≥98.0% (GC)

Methyl heneicosanoate

~99% (capillary GC), crystalline

Nonadecanoic acid

≥98% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?