Synonym:
4,4′-(1,2-Diethylethylene)diphenol,
meso-
CAS Number 84-16-2
Empirical Formula (Hill Notation) C18H22O2
Molecular Weight 270.37
Beilstein Registry Number 3209460
EC Number 201-518-1
MDL number MFCD00068996
PubChem Substance ID 24895805
Popular Documents: Specification Sheet (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Bioactive Small Molecules, Cell Biology, |
| grade | analytical standard |
| assay | ≥98% |
| Gene Information |
human ... ESR1(2099) rat ... Esr1(24890) |
The carcinogen hexestrol is a nonsteroidal synthetic estrogen. Metabolic activation of hexestrol to its quinone, which reacts with DNA to form analogous depurinating adducts, may be a primary critical event leading to oncogenic mutations and cancer initiation.
Reduced analogue of diethylstilbestrol.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?