IRMM541 Fluka
Oxide glass
BCR® certified Reference Material
DOWNLOAD MSDS (PDF)
Description
Analysis Note
uranium mass fraction: ~55 mg U/kg
Physical form
discs, 15 mm diameter
Legal Information
BCR is a registered trademark of European Commission
Price and Availability
Customers Also Viewed
Thioglycolic acid solution
~80% in H2O, for spectrophotometric det. of palladium, iron, uranium(VI), molybdates and nitrites
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?