Synonym:
1-
CAS Number 68813-55-8
Empirical Formula (Hill Notation) C13H16N2O · C23H16O6
Molecular Weight 604.65
EC Number 272-332-6
MDL number MFCD19105662
PubChem Substance ID 24898003
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Antibiotics, Antibiotics A to Z, |
| grade | analytical standard |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?