PHR1008 Fluka

L-Ascorbic acid

pharmaceutical secondary standard; traceable to USP, PhEur and BP

DOWNLOAD MSDS (PDF)

Synonym: L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

  • CAS Number 50-81-7

  • Empirical Formula (Hill Notation) C6H8O6

  • Molecular Weight 176.12

  •  Beilstein Registry Number 84272

  •  E Number E300

  •  EC Number 200-066-2

  •  MDL number MFCD00064328

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Chromatography, E numbers (Food Additives), E300,
grade   certified reference material
packaging   pkg of 2 g
mp   190-194 °C (dec.)
pharmacopeia traceability   traceable to BP 461
  traceable to PhEur A1300000
  traceable to USP 1043003
storage temp.   2-8°C

Price and Availability

Customers Also Viewed

<SC>L</SC>-Ascorbic acid

BioXtra, ≥99.0%, crystalline

<SC>L</SC>-Ascorbic acid

analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?