Synonym: Vitamin E acetate, all-rac-α-Tocopheryl acetate
CAS Number 7695-91-2
Empirical Formula (Hill Notation) C31H52O3
Molecular Weight 472.74
Beilstein Registry Number 97512
EC Number 231-710-0
MDL number MFCD00072042
| Related Categories | Analytical Standards, Analytical/Chromatography, Chromatography, Pharmaceutical Secondary Standards, Pharmaceutical Standards More... |
| grade | certified reference material |
| packaging | pkg of 500 mg |
| density | 0.96 g/mL at 20 °C(lit.) |
| pharmacopeia traceability | traceable to PhEur T1600000 |
| traceable to USP 1667701 |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?