Synonym:
6-
CAS Number 58-93-5
Empirical Formula (Hill Notation) C7H8ClN3O4S2
Molecular Weight 297.74
EC Number 200-403-3
MDL number MFCD00051765
| Related Categories | All Doping Standards (A-Z), Analytical Standards, Analytical/Chromatography, Chromatography, Diuretic, |
| grade | certified reference material |
| packaging | pkg of 1 g |
| pharmacopeia traceability | traceable to BP 186 |
| traceable to PhEur H1200000 | |
| traceable to USP 1314009 |
Carbonic anhydrase inhibitor
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?