PHR1113 Fluka

Sodium L-lactate

pharmaceutical secondary standard; traceable to USP

DOWNLOAD MSDS (PDF)

Synonym: (S)-2-Hydroxypropionic acid sodium salt, L-Lactic acid sodium salt, Sarcolactic acid sodium salt

  • CAS Number 867-56-1

  • Empirical Formula (Hill Notation) C3H5NaO3

  • Molecular Weight 112.06

  •  Beilstein Registry Number 4567087

  •  E Number E325

  •  EC Number 212-762-3

  •  MDL number MFCD00066576

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Chromatography, E numbers (Food Additives), E300–E399 (antioxidants, acidity regulators),
grade   certified reference material
packaging   pkg of 1 g
mp   163-165 °C(lit.)
pharmacopeia traceability   traceable to USP 1614308

Price and Availability

Customers Also Viewed

TraceCERT®, 1000 mg/L lactate in H2O

Sodium <SC>D</SC>-lactate

≥99.0% (NT)

Sodium <SC>DL</SC>-lactate

ReagentPlus®, ≥99.0% (NT)

Sodium <SC>L</SC>-lactate

≥99.0% (NT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?