Synonym: 2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC
CAS Number 128-37-0
Linear Formula [(CH3)3C]2C6H2(CH3)OH
Molecular Weight 220.35
Beilstein Registry Number 1911640
E Number E321
EC Number 204-881-4
MDL number MFCD00011644
| Related Categories | Analytical Standards, Analytical/Chromatography, Chromatography, E numbers (Food Additives), E300–E399 (antioxidants, acidity regulators), |
| grade | certified reference material |
| vapor density | 7.6 (vs air) |
| vapor pressure | <0.01 mmHg ( 20 °C) |
| autoignition temp. | 878 °F |
| packaging | pkg of 1 g |
| bp | 265 °C(lit.) |
| mp | 69-73 °C(lit.) |
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO Guide 34.
Values of analytes vary lot to lot.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?