CAS Number 1218-34-4
Empirical Formula (Hill Notation) C13H14N2O3
Molecular Weight 246.26
MDL number MFCD00065976
| Related Categories | Analytical Standards, Analytical/Chromatography, Chromatography, Pharmaceutical Secondary Standards, Pharmaceutical Standards More... |
| grade | certified reference material |
| packaging | pkg of 500 mg |
| pharmacopeia traceability | traceable to USP 1700523 |
| storage temp. | 2-8°C |
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO Guide 34.
Values of analytes vary lot to lot.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?