Synonym:
2-
CAS Number 154-69-8
Empirical Formula (Hill Notation) C16H21N3 · HCl
Molecular Weight 291.82
EC Number 205-833-5
MDL number MFCD00012815
PubChem Substance ID 24900456
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Synthesis, Chromatography, |
| grade | analytical standard, for drug analysis |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?