EMAIL THIS PAGE TO A FRIEND

09830 Fluka

Ammonium bicarbonate

BioUltra, ≥99.5% (T)

Synonym: Ammonium hydrogen carbonate

Purchase

Properties

Related Categories Alphabetical, BioUltra Buffers, Biochemicals and Reagents, Biological Buffers
vapor density   2.7 (vs air)
product line   BioUltra
assay   ≥99.5% (T)
impurities   insoluble matter, passes filter test
pH   7.0-8.5 (25 °C, 1 M in H2O)
solubility   H2O: soluble1 M at 20 °C, clear, colorless
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Al: ≤5 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤500 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Zn: ≤5 mg/kg
λ   1 M in H2O
UV absorption   λ: 260 nm Amax: ≤0.030
  λ: 280 nm Amax: ≤0.020

Description

Application

Do you have application information on this product that you would like to share?

Ammonium bicarbonate can be used to study biological buffers. Ammonium bicarbonate has been used in a study that demonstrated that ammonium bicarbonate salts, which can be regenerated using low-temperature waste heat, can also produce sufficient voltage for hydrogen gas generation in a microbial reverse-electrodialysis electrolysis cells. Ammonium bicarbonate has also been used in a study that developed a fast and sensitive method for the simultaneous determination of Sudan dyes in food samples using partial filling micellar electrokinectic chromatography-mass spectrometry.

Other Notes

Sample loading solution for the gas-phase sequenator1; Removable eluant in ion-exchange chromatography2; Buffer component for the digestion of tRNA3.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



TruPAGE™ Pre-cast Polyacrylamide Gels


TruPAGE™ Pre-cast Polyacrylamide GelsTruPAGE Gels run true every time!

  • . 2-Year Shelf Life
  • . Superior Band Resolution
  • . Strong & Tear Resistant
  • . Higher Capacity Wells


Customers Also Viewed

Ammonium bicarbonate

ReagentPlus®, ≥99.0%

Ammonium bicarbonate

eluent additive for LC-MS

Ammonium bicarbonate

puriss., meets analytical specification of Ph.Eur., BP, E 503, 99-101%

Ammonium carbonate

99.999% trace metals basis

Ammonium carbonate

ACS reagent, ≥30.0% NH3 basis

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
Hazard Codes (Europe) 
Xn
Risk Statements (Europe) 
22
WGK Germany 
1
RTECS 
BO8600000

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. M.W. Hunkapiller et al. Meth. Enzymol. 91, 402, (1983)

2. R.J. Carrico et al. Meth. Enzymol. 57, 117, (1979)

3. P.R. Schimmel, H.J.P. Shoemaker Meth. Enzymol. 59, 343, (1979)

Electrothermal supercharging of proteins in native electrospray ionization Sterling, H., et al. Anal. Chem. 84, 3795-801, (2012)

Loading...


Proteomic identification of FHL1 as the protein mutated in human reducing body myopathy Judkins, A.R., et al. J. Clin. Invest. 118, 904-912, (2008)

Loading...


Hydrogen generation in microbial reverse-electrodialysis electrolysis cells using a heat-regenerated salt solution Nam , J., et al. Environ. Sci. Technol. 46, 5240-6, (2012)

Loading...


Disruption of the arsenic (+3 oxidation state) methyltransferase gene in the mouse alters the phenotype for methylation of arsenic and affects distribution and retention of orally administered arsenate. Drobna Z Chem. Res. Toxicol. 22(10), 1713-20, (2009)

Loading...


Analysis of ammonia-oxidizing bacteria dominating in lab-scale bioreactors with high ammonium bicarbonate loading. Vejmelkova D, Sorokin DY, Abbas B, et al. Appl. Microbiol. Biotechnol. 93(1), 401-10, (2012)

Loading...


Development of different analysis platforms with LC-MS for pharmacokinetic studies of protein drugs. Lu Q Anal. Chem. 81(21), 8715-23, (2009)

Loading...


High throughput proteomic analysis of the secretome in an explant model of articular cartilage inflammation. Abigail L. Clutterbuck et al J. Proteomics 74, 704-15, (2011)

Loading...


Quantifying protein interface footprinting by hydroxyl radical oxidation and molecular dynamics simulation: application to galectin-1. Charvátová O J. Am. Soc. Mass Spectrom. 19(11), 1692-705, (2008)

Loading...


Acrylamide formation and antioxidant level in biscuits related to recipe and baking. Haase NU, Grothe KH, Matthäus B, et al. Food Addit. Contam. Part A. Chem. Anal. Control. Expo. Risk Assess. 29(8), 1230-8, (2012)

Loading...


Urine as a CO2 absorbent. Aguilar MJ J. Hazard. Mater. 213-214, 502-4, (2012)

Loading...


Preparation and characterization of novel poly(ε-caprolactone)/biphasic calcium phosphate hybrid composite microspheres. Bao TQ, Franco RA, and Lee BT J. Biomed. Mater. Res. B. Appl. Biomater. 98(2), 272-9, (2011)

Loading...


[Biological activity evaluation of porous HA ceramics using NH4 HCO3/PVA as pore-creating agents]. Wang S and Zhang D Sheng Wu Yi Xue Gong Cheng Xue Za Zhi 27(6), 1280-5, (2010)

Loading...


[Influence of land-use type on moisture-effect of nitrification in subtropical red soils]. Qian C and Cai ZC Huan Jing Ke Xue 31(10), 2417-22, (2010)

Loading...


Preparation of budesonide-loaded porous PLGA microparticles and their therapeutic efficacy in a murine asthma model. Oh YJ, Lee J, Seo JY, et al. J. Control. Release 150(1), 56-62, (2011)

Loading...


Two-step stacking in capillary zone electrophoresis featuring sweeping and micelle to solvent stacking: II. Organic anions. Quirino JP and Guidote AM J. Chromatogr. A 1218(7), 1004-10, (2011)

Loading...


Metal ions and electrolytes regulate the dissociation of heme from human hemopexin at physiological pH. Mauk MR and Mauk AG J. Biol. Chem. 285(27), 20499-506, (2010)

Loading...


Energy capture from thermolytic solutions in microbial reverse-electrodialysis cells. Cusick RD, Kim Y, and Logan BE Science 335(6075), 1474-7, (2012)

Loading...


On-line sample concentration of organic anions in capillary zone electrophoresis by micelle to solvent stacking. Guidote AM and Quirino JP J. Chromatogr. A 1217(40), 6290-5, (2010)

Loading...


The palladium-catalyzed cyanation of indole C-H bonds with the combination of NH4HCO3 and DMSO as a safe cyanide source. Ren, X.; et al. Chem. Commun. (Camb.) 47, 6725, (2011)

Loading...


A protocol for the production of recombinant spider silk-like proteins for artificial fiber spinning. Florence Teulé et al Nat. Protoc. 4, 341-55, (2009)

Loading...


Formulation and in vivo evaluation of sodium alendronate spray-dried microparticles intended for lung delivery. Cruz L, Fattal E, Tasso L, et al. J. Control. Release 152(3), 370-5, (2011)

Loading...


On-plate selective enrichment and self-desalting of peptides/proteins for direct MALDI MS analysis. Zeng Z, Wang Y, Shi S, et al. Anal. Chem. 84(5), 2118-23, (2012)

Loading...


Electrokinetic supercharging-electrospray ionisation-mass spectrometry for separation and on-line preconcentration of hypolipidaemic drugs in water samples. Dawod M, Breadmore MC, Guijt RM, et al. Electrophoresis 31(7), 1184-93, (2010)

Loading...


Histo-blood group antigens act as attachment factors of rabbit hemorrhagic disease virus infection in a virus strain-dependent manner. Nyström K PLoS Pathog. 7(8), e1002188, (2011)

Loading...


A modified lead-matrix separation procedure shown for lead isotope analysis in Trojan silver artefacts as an example. Vogl J, Paz B, Koenig M, et al. Anal. Bioanal. Chem 405(9), 2995-3000, (2013)

Loading...


Experimental study of a 4040 spiral-wound forward-osmosis membrane module. Kim YC and Park SJ Environ. Sci. Technol. 45(18), 7737-45, (2011)

Loading...


The impact of lower gut nitrogen supply on nitrogen balance and urea kinetics in growing pigs fed a valine-limiting diet. Columbus D, Lapierre H, Fuller MF, et al. J. Anim. Sci. 90 Suppl 4, 62-4, (2012)

Loading...


Evaluation of yellow rectangle traps coated with hot melt pressure sensitive adhesive and sticky gel against Rhagoletis indifferens (Diptera: Tephritidae). Yee WL J. Econ. Entomol. 104(3), 909-19, (2011)

Loading...


Sensitive and fast determination of Sudan dyes in chili powder by partial-filling micellar electrokinetic chromatography-tandem mass spectrometry Fukuji, T., et al. Electrophoresis 33, 705-12, (2012)

Loading...


LC-MS/MS method using unbonded silica column and aqueous/methanol mobile phase for the simultaneous quantification of a drug candidate and co-administered metformin in rat plasma. Discenza L, D'Arienzo C, Olah T, et al. J. Chromatogr. B. Analyt. Technol. Biomed. Life Sci. 878(19), 1583-9, (2010)

Loading...


On-line sample concentration and determination of cationic alkaloids in human plasma by micelle to solvent stacking in capillary zone electrophoresis. Zhang S, Ma R, Yang X, et al. J. Chromatogr. B. Analyt. Technol. Biomed. Life Sci. 906, 41-7, (2012)

Loading...


Emissions of NO and NH3 from a typical vegetable-land soil after the application of chemical N fertilizers in the Pearl River Delta. Li D PLoS ONE 8(3), e59360, (2013)

Loading...


Intracellular adaptation of Brucella abortus. Lamontagne J J. Proteome Res. 8(3), 1594-609, (2009)

Loading...


Preparation and in vitro evaluation of poly(D,L-lactide-co-glycolide) air-filled nanocapsules as a contrast agent for ultrasound imaging. Néstor MM, Kei NP, Guadalupe NA, et al. Ultrasonics 51(7), 839-45, (2011)

Loading...


Formulation of pyrazinamide-loaded large porous particles for the pulmonary route: avoiding crystal growth using excipients. Pham DD, Fattal E, Ghermani N, et al. Int. J. Pharm. 454(2), 668-77, (2013)

Loading...


Efficacy of wax matrix bait stations for Mediterranean fruit flies (Diptera: Tephritidae). Epsky ND, Midgarden D, Rendón P, et al. J. Econ. Entomol. 105(2), 471-9, (2012)

Loading...


An optical-density-based feedback feeding method for ammonium concentration control in Spirulina platensis cultivation. Bao Y, Wen S, Cong W, et al. J. Microbiol. Biotechnol. 22(7), 967-74, (2012)

Loading...


Merck 14,497

Corp MSDS 1 (1), 232:C / FT-IR 2 (3), 4733:A / RegBook 1 (3), 3401:M / Sigma FT-IR 1 (2), 1012:A

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?