EMAIL THIS PAGE TO A FRIEND

45315 Fluka

Acephate

PESTANAL®, analytical standard

Synonym: O,S-Dimethyl N-acetylphosphoramidothioate

  • CAS Number 30560-19-1

  • Empirical Formula (Hill Notation) C4H10NO3PS

  • Molecular Weight 183.17

  •  Beilstein Registry Number 1936365

  •  EC Number 250-241-2

  •  MDL number MFCD00055361

  •  PubChem Substance ID 24868762

Purchase

Description

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

QuEChERS SPE Tubes


QuEChERS SPE TubesSupelco provides a range of routine and specialty products for this popular sample prep technique used in multi-pesticide residue analysis.




Solvents for Pesticide Residue Analysis


Solvents for Pesticide Residue AnalysisFluka solvents for residue analysis of pesticides are glass-distilled and
extensively tested by GC/ECD to ensure minimum halogenated residue.




Customers Also Viewed

Acephate

analytical standard

Azinphos-ethyl

PESTANAL®, analytical standard

Methamidophos

PESTANAL®, analytical standard

Omethoate

PESTANAL®, analytical standard

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
Hazard Codes (Europe) 
Xn
Risk Statements (Europe) 
22
Safety Statements (Europe) 
36
WGK Germany 
2
RTECS 
TB4760000

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Repeated pesticide exposure among North Carolina migrant and seasonal farmworkers. Arcury TA, Grzywacz JG, Talton JW, et al. Am. J. Ind. Med. 53(8), 802-13, (2010)

Loading...


Degradation and mineralization kinetics of acephate in humid tropic soils of Malaysia. Chai LK, Wong MH, Mohd-Tahir N, et al. Chemosphere 79(4), 434-40, (2010)

Loading...


Effect of repeated applications of buprofezin and acephate on soil cellulases, amylase, and invertase. Raju MN and Venkateswarlu K Environ. Monit. Assess. 186(10), 6319-25, (2014)

Loading...


Acetylcholinesterase based biosensor for monitoring of Malathion and Acephate in food samples: a voltammetric study. Raghu P, Madhusudana Reddy T, Reddaiah K, et al. Food Chem. 142, 188-96, (2014)

Loading...


Inhibition of cholinesterase activity by soil extracts and predicted environmental concentrations (PEC) to select relevant pesticides in polluted soils. Meza JC, Perez PA, Salin MB, et al. J. Environ. Sci. Health B 45(3), 214-21, (2010)

Loading...


Sol-gel immobilized biosensor for the detection of organophosphorous pesticides: a voltammetric method. Raghu P, Swamy BE, Reddy TM, et al. Bioelectrochemistry 83, 19-24, (2012)

Loading...


Ultrasensitively sensing acephate using molecular imprinting techniques on a surface plasmon resonance sensor. Wei C, Zhou H, and Zhou J Talanta 83(5), 1422-7, (2011)

Loading...


Relative influence of plant quality and natural enemies on the seasonal dynamics of Bemisia tabaci (Hemiptera: Aleyrodidae) in cotton. Asiimwe P, Naranjo SE, and Ellsworth PC J. Econ. Entomol. 106(3), 1260-73, (2013)

Loading...


Acephate resistance in populations of the tarnished plant bug (Heteroptera: Miridae) from the Mississippi River Delta. Snodgrass GL, Gore J, Abel CA, et al. J. Econ. Entomol. 102(2), 699-707, (2009)

Loading...


Characterization of acetylcholinesterase in Hong Kong oyster (Crassostrea hongkongensis) from South China Sea. Zha G, Chen VP, Luk WK, et al. Chem. Biol. Interact. 203(1), 277-81, (2013)

Loading...


Metabolomic analysis of the toxic effect of chronic low-dose exposure to acephate on rats using ultra-performance liquid chromatography/mass spectrometry. Hao DF, Xu W, Wang H, et al. Ecotoxicol. Environ. Saf. 83, 25-33, (2012)

Loading...


Influence of polyphenolic extracts from Enydra fluctuans on oxidative stress induced by acephate in rats. Datta S, Dhar P, Mukherjee A, et al. Food Chem. Toxicol. 48(10), 2766-71, (2010)

Loading...


Mineralization of acephate, a recalcitrant organophosphate insecticide is initiated by a pseudomonad in environmental samples. Pinjari AB, Novikov B, Rezenom YH, et al. PLoS ONE 7(4), e31963, (2012)

Loading...


Esterase-based resistance in the tobacco-adapted form of the green peach aphid, Myzus persicae (Sulzer) (Hemiptera: Aphididae) in the eastern United States. Srigiriraju L, Semtner PJ, Anderson TD, et al. Arch. Insect Biochem. Physiol. 72(2), 105-23, (2009)

Loading...


Insecticide susceptibility of Euschistus heros (Heteroptera: Pentatomidae) in Brazil. Sosa-Gómez DR, Da Silva JJ, Lopes Ide O, et al. J. Econ. Entomol. 102(3), 1209-16, (2009)

Loading...


In vitro induction/inhibition of germinal vesicle breakdown (GVBD) in frog (Euphlyctis cyanophlyctis) oocytes by endocrine active compounds. Ghodageri MG and Katti P Drug Chem. Toxicol. 36(2), 217-23, (2013)

Loading...


Residue analysis of acephate and its metabolite methamidophos in open field and greenhouse pakchoi (Brassica campestris L.) by gas chromatography-tandem mass spectrometry. Chuanjiang T, Dahui L, Xinzhong Z, et al. Environ. Monit. Assess. 165(1-4), 685-92, (2010)

Loading...


Effects of waterborne acephate exposure on antioxidant responses and acetylcholinesterase activities in Synechogobius hasta. Liu XJ, Luo Z, Zheng JL, et al. Environ. Toxicol. 28(1), 42-50, (2013)

Loading...


Reduction of pesticide residues on fresh vegetables with electrolyzed water treatment. Hao J, Wuyundalai, Liu H, et al. J. Food Sci. 76(4), C520-4, (2011)

Loading...


Microarray analysis of gene regulations and potential association with acephate-resistance and fitness cost in Lygus lineolaris. Zhu YC, Guo Z, He Y, et al. PLoS ONE 7(5), e37586, (2012)

Loading...


Residues of acephate and its metabolite methamidophos in/on mango fruit (Mangifera indica L.). Mohapatra S, Ahuja AK, Deepa M, et al. Bull. Environ. Contam. Toxicol. 86(1), 101-4, (2011)

Loading...


Acephate immunotoxicity in White Leghorn cockerel chicks upon experimental exposure. Tripathi SM, Thaker AM, Joshi CG, et al. Environ. Toxicol. Pharmacol. 34(2), 192-9, (2012)

Loading...


[Analysis of acephate, methamidophos and omethoate in animal and fishery products, and honey by LC-MS]. Ueno E, Ohno H, Tanahashi T, et al. Shokuhin Eiseigaku Zasshi 51(3), 122-7, (2010)

Loading...


Controlled release formulations of acephate: water and soil release kinetics. Nisar K, Kumar J, Shakil NA, et al. J. Environ. Sci. Health B 44(6), 533-7, (2009)

Loading...


Entomotoxicity and biosafety assessment of PEGylated acephate nanoparticles: a biologically safe alternative to neurotoxic pesticides. Pradhan S, Roy I, Lodh G, et al. J. Environ. Sci. Health B 48(7), 559-69, (2013)

Loading...


[Development of a novel multi-functional adsorbent bearing with long-chain hydrophobic and anion exchange groups for the simple and rapid determination of residual acephate in vegetables]. Tsukamoto T, Shimizu M, Yamamoto A, et al. Shokuhin Eiseigaku Zasshi 51(2), 58-64, (2010)

Loading...


Concentration-mortality responses of Myzus persicae and natural enemies to selected insecticides. Bacci L, Rosado JF, Picanço MC, et al. J. Environ. Sci. Health. A. Tox. Hazard. Subst. Environ. Eng. 47(12), 1930-7, (2012)

Loading...


Calculation of the dietary exposure of Chinese consumers to acephate residues using deterministic and probabilistic approaches. Liu P, Li CY, Wang CN, et al. Food Addit. Contam. Part A. Chem. Anal. Control. Expo. Risk Assess. 28(5), 649-58, (2011)

Loading...


Merck 14,31

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?