EMAIL THIS PAGE TO A FRIEND

45449 Fluka

Dimethoate

PESTANAL®, analytical standard

  • CAS Number 60-51-5

  • Empirical Formula (Hill Notation) C5H12NO3PS2

  • Molecular Weight 229.26

  •  Beilstein Registry Number 1785339

  •  EC Number 200-480-3

  •  MDL number MFCD00053676

  •  PubChem Substance ID 24868923

Purchase

Description

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

QuEChERS SPE Tubes


QuEChERS SPE TubesSupelco provides a range of routine and specialty products for this popular sample prep technique used in multi-pesticide residue analysis.




Solvents for Pesticide Residue Analysis


Solvents for Pesticide Residue AnalysisFluka solvents for residue analysis of pesticides are glass-distilled and
extensively tested by GC/ECD to ensure minimum halogenated residue.




Customers Also Viewed

Dimethoate

analytical standard

Methamidophos

PESTANAL®, analytical standard

Omethoate

PESTANAL®, analytical standard

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
Hazard Codes (Europe) 
Xn
Risk Statements (Europe) 
Safety Statements (Europe) 
36/37
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
TE1750000
Flash Point(F) 
224.6 °F
Flash Point(C) 
107 °C

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Biomarkers and energy reserves in the isopod Porcellionides pruinosus: the effects of long-term exposure to dimethoate. Ferreira NG, Morgado R, Santos MJ, et al. Sci. Total Environ. 502, 91-102, (2015)

Loading...


Changes of drug metabolizing enzymes in the liver of male sheep exposed to either cypermethrin or dimethoate. Sheweita SA, Yousef MI, Baghdadi HH, et al. Drug Metab. Lett. 6(1), 2-6, (2012)

Loading...


Gene expression responses linked to reproduction effect concentrations (EC 10,20,50,90) of dimethoate, atrazine and carbendazim, in Enchytraeus albidus. Novais SC, De Coen W, and Amorim MJ PLoS ONE 7(4), e36068, (2012)

Loading...


Computational and experimental investigation of molecular imprinted polymers for selective extraction of dimethoate and its metabolite omethoate from olive oil. Bakas I, Oujji NB, Moczko E, et al. J. Chromatogr. A 1274, 13-8, (2013)

Loading...


Additive effects of predator cues and dimethoate on different levels of biological organisation in the non-biting midge Chironomus riparius. Van Praet N, De Jonge M, Stoks R, et al. Aquat. Toxicol. 155, 236-43, (2014)

Loading...


A role for solvents in the toxicity of agricultural organophosphorus pesticides. Eddleston M, Street JM, Self I, et al. Toxicology 294(2-3), 94-103, (2012)

Loading...


Species-specific differences in biomarker responses in two ecologically different earthworms exposed to the insecticide dimethoate. Velki M and Hackenberger BK Comp. Biochem. Physiol. C. Toxicol. Pharmacol. 156(2), 104-12, (2012)

Loading...


Chloride anion effect on the advanced oxidation processes of methidathion and dimethoate: role of Cl2(·-) radical. Caregnato P, Rosso JA, Soler JM, et al. Water Res. 47(1), 351-62, (2013)

Loading...


Protective effects of vitamin E and selenium against dimethoate-induced cardiotoxicity in vivo: biochemical and histological studies. Amara IB, Soudani N, Hakim A, et al. Environ. Toxicol. 28(11), 630-43, (2013)

Loading...


Acetylcholinesterase immobilized on magnetic beads for pesticides detection: application to olive oil analysis. Ben Oujji N, Bakas I, Istamboulié G, et al. Sensors (Basel.) 12(6), 7893-904, (2012)

Loading...


Sub-chronic exposure to the insecticide dimethoate induces a proinflammatory status and enhances the neuroinflammatory response to bacterial lypopolysaccharide in the hippocampus and striatum of male mice. Astiz M, Diz-Chaves Y, and Garcia-Segura LM Toxicol. Appl. Pharmacol. 272(2), 263-71, (2013)

Loading...


Extensive Ace2 duplication and multiple mutations on Ace1 and Ace2 are related with high level of organophosphates resistance in Aphis gossypii. Shang Q, Pan Y, Fang K, et al. Environ. Toxicol. 29(5), 526-33, (2014)

Loading...


Antioxidative responses in females and males of the spider Xerolycosa nemoralis (Lycosidae) exposed to natural and anthropogenic stressors. Wilczek G, Babczyńska A, and Wilczek P Comp. Biochem. Physiol. C. Toxicol. Pharmacol. 157(2), 119-31, (2013)

Loading...


Dimethoate affects cholinesterases in Folsomia candida and their locomotion--false negative results of an avoidance behaviour test. Pereira CM, Novais SC, Soares AM, et al. Sci. Total Environ. 443, 821-7, (2013)

Loading...


Factors affecting soil residues of dieldrin, endosulfan, gamma-HCH, dimethoate, and pyrolan. El Beit IO, Wheelock JV, and Cotton DE Ecotoxicol. Environ. Saf. 5(2), 135-60, (1981)

Loading...


A new method for analysis of the toxicity of organophosphorus pesticide, dimethoate on rotifer based on response surface methodology. Guo R, Ren X, and Ren H J. Hazard. Mater. 237-238, 270-6, (2012)

Loading...


Alteration of leaf metabolism in Bt-transgenic rice (Oryza sativa L.) and its wild type under insecticide stress. Zhou J, Zhang L, Chang Y, et al. J. Proteome Res. 11(8), 4351-60, (2012)

Loading...


Maternal exposure to the mixture of organophosphorus pesticides induces reproductive dysfunction in the offspring. Yu Y, Yang A, Zhang J, et al. Environ. Toxicol. 28(9), 507-15, (2013)

Loading...


Nucleophilic substitution reaction at the nitrogen of arylsulfonamides with phosphide anion. Yoshida S, Igawa K, and Tomooka K J. Am. Chem. Soc. 134(47), 19358-61, (2012)

Loading...


Can nutrients mask community responses to insecticide mixtures? Alexander AC, Luis AT, Culp JM, et al. Ecotoxicology 22(7), 1085-100, (2013)

Loading...


Metabonomics analysis of urine and plasma from rats given long-term and low-dose dimethoate by ultra-performance liquid chromatography-mass spectrometry. Feng Z, Sun X, Yang J, et al. Chem. Biol. Interact. 199(3), 143-53, (2012)

Loading...


A new OECD test guideline for the predatory soil mite Hypoaspis aculeifer: results of an international ring test. Smit CE, Moser T, and Römbke J Ecotoxicol. Environ. Saf. 82, 56-62, (2012)

Loading...


Variation in effects of four OP insecticides on photosynthetic pigment fluorescence of Chlorella vulgaris Beij. Jena S, Acharya S, and Mohapatra PK Ecotoxicol. Environ. Saf. 80, 111-7, (2012)

Loading...


Influence of organophosphate poisoning on human dendritic cells. Schäfer M, Koppe F, Stenger B, et al. Chem. Biol. Interact. 206(3), 472-8, (2013)

Loading...


Single versus combined lethal effects of three agricultural insecticides on larvae of the freshwater insect Chironomus dilutus. LeBlanc HM, Culp JM, Baird DJ, et al. Arch. Environ. Contam. Toxicol. 63(3), 378-90, (2012)

Loading...


Effects of solvent on the toxicity of dimethoate in a honey bee in vitro larval study. Wilkins S, Jarratt N, Harkin S, et al. Pest Manag. Sci. 69(4), 462-3, (2013)

Loading...


Baseline susceptibility of Planococcus ficus (Hemiptera: Pseudococcidae) from California to select insecticides. Prabhaker N, Gispert C, and Castle SJ J. Econ. Entomol. 105(4), 1392-400, (2012)

Loading...


Does insecticide drift adversely affect grasshoppers (Orthoptera: Saltatoria) in field margins? A case study combining laboratory acute toxicity testing with field monitoring data. Bundschuh R, Schmitz J, Bundschuh M, et al. Environ. Toxicol. Chem. 31(8), 1874-9, (2012)

Loading...


Assessment the toxic effects of dimethoate to rotifer using swimming behavior. Guo R, Ren X, and Ren H Bull. Environ. Contam. Toxicol. 89(3), 568-71, (2012)

Loading...


Kinetics and mechanism for omethoate degradation by catalytic ozonation with Fe(III)-loaded activated carbon in water. Qiang Z, Ling W, and Tian F Chemosphere 90(6), 1966-72, (2013)

Loading...


Reduction of pesticide residues on fresh vegetables with electrolyzed water treatment. Hao J, Wuyundalai, Liu H, et al. J. Food Sci. 76(4), C520-4, (2011)

Loading...


Exogenous arachidonate restores the dimethoate-induced inhibition of steroidogenesis in rat interstitial cells. Astiz M, Hurtado de Catalfo G, de Alaniz MJ, et al. Lipids 47(6), 557-69, (2012)

Loading...


Activity of selected antioxidative enzymes in rats exposed to dimethoate and pyrantel tartrate. Barski D and Spodniewska A Pol. J. Vet. Sci. 15(2), 239-45, (2012)

Loading...


Concentration-mortality responses of Myzus persicae and natural enemies to selected insecticides. Bacci L, Rosado JF, Picanço MC, et al. J. Environ. Sci. Health. A. Tox. Hazard. Subst. Environ. Eng. 47(12), 1930-7, (2012)

Loading...


Monitoring of pesticide residues levels in fresh vegetable form Heibei Province, North China. Li W, Tai L, Liu J, et al. Environ. Monit. Assess. 186(10), 6341-9, (2014)

Loading...


Dimethoate induced oxidative damage and histopathological changes in lung of adult rats: modulatory effects of selenium and/or vitamin E. Amara IB, Soudani N, Troudi A, et al. Biomed. Environ. Sci. 25(3), 340-51, (2012)

Loading...


Carcinogenicity of dimethoate. Reuber MD Environ. Res. 34(2), 193-211, (1984)

Loading...


Merck 14,3220

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?