EMAIL THIS PAGE TO A FRIEND

77608 Fluka

Phenol

BioUltra, for molecular biology, ≥99.5% (GC)

Synonym: Hydroxybenzene

Properties

Related Categories Acids and Bases, Alphabetical, BioUltra, Biochemicals and Reagents, DNA & RNA Purification,
grade   for molecular biology
vapor density   3.24 (vs air)
vapor pressure   0.09 psi ( 55 °C)
  0.36 mmHg ( 20 °C)
product line   BioUltra
assay   ≥99.5% (GC)
autoignition temp.   1319 °F
contains   ~0.15% hypophosphorous acid as stabilizer
expl. lim.   8.6 %
impurities   DNases, none detected
  RNases, none detected
  insoluble matter, passes filter test
  phosphatases, none detected
  proteases, none detected
  ≤0.5% water
ign. residue   ≤0.05%
pH   3.0-6.0 (25 °C, 0.5 M in H2O)
bp   182 °C(lit.)
mp   40-42 °C(lit.)
  40-42 °C
solubility   H2O: soluble0.5 M at 20 °C, clear, colorless
density   1.071 g/mL at 25 °C(lit.)
anion traces   chloride (Cl-): ≤5 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Al: ≤5 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Zn: ≤5 mg/kg
absorption   absorption0.5M/294 nm, H2O cut-off
storage temp.   2-8°C

Description

Other Notes

For the preparation of DNA 1; Large- and small-scale phenol extractions2

Price and Availability


LifeSep Magnetic Separator

Genomics Promo

RNAi Offer

Customers Also Viewed

Phenol
77607Phenol

BioUltra, for molecular biology, TE-saturated, ~73% (T)

Phenol
77613Phenol

for molecular biology, ~90% (T), liquid

Phenol
35952Phenol

PESTANAL®, analytical standard

Phenol
77610Phenol

puriss. p.a., ACS reagent, ≥99.5% (GC), crystalline (loose)

Phenol
W322340Phenol

natural, 97%, FG

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?