CAS Number 7681-49-4
Linear Formula NaF
Molecular Weight 41.99
EC Number 231-667-8
MDL number MFCD00003524
PubChem Substance ID 24845145
| Related Categories | Chemical Synthesis, Inorganic Salts, Sodium, Synthetic Reagents |
| vapor pressure | 1.4 mmHg ( 0 °C) |
| grade | puriss. |
| assay | 98.5-100.5% (calc. to the dried substance) |
| quality | meets analytical specification of Ph. Eur., BP, USP |
| impurities | acidity or alkalinity, complies |
| organic volatile impurities, complies (GC) | |
| residual solvents, complies |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?